Assay Details
| Gene Symbol: lin-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029416.4 | NP_001024587.1 | 6 - 7 | 741 | 94 | |
| GenBank mRNA | X92564.1 | - | 725 | 94 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

lin-2| Gene Symbol: lin-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029416.4 | NP_001024587.1 | 6 - 7 | 741 | 94 | |
| GenBank mRNA | X92564.1 | - | 725 | 94 | ||
Set Membership: |
|