Assay Details
| Gene Symbol: adt-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001375156.2 | NP_001361951.1 | 16 - 17 | 2724 | 92 | |
| NM_077715.5 | NP_510116.2 | 16 - 17 | 2733 | 92 | ||
| GenBank mRNA | AB066246.1 | - | 16 - 17 | 2733 | 92 | |
More Information
Set Membership: |
|

adt-1
Genomic Map