Assay Details
| Gene Symbol: adt-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001375156.2 | NP_001361951.1 | 26 - 27 | 4449 | 82 | |
| NM_077715.5 | NP_510116.2 | 26 - 27 | 4458 | 82 | ||
| GenBank mRNA | AB066246.1 | - | 26 - 27 | 4458 | 82 | |
More Information
Set Membership: |
|

adt-1
Genomic Map