Assay Details
Gene Symbol: rgs-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077723.6 | NP_510124.2 | 1 - 2 | 320 | 74 | |
GenBank mRNA | AF220265.1 | - | 1 - 2 | 283 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
