Assay Details
Gene Symbol: rgs-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129662.4 | NP_001123134.1 | 2 - 3 | 393 | 77 | |
NM_001270270.5 | NP_001257199.1 | 2 - 3 | 579 | 77 | ||
NM_001383658.1 | 2 - 3 | 143 | 77 | |||
NM_077723.6 | NP_510124.2 | 2 - 3 | 365 | 77 | ||
GenBank mRNA | AF220265.1 | - | 2 - 3 | 328 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
