Assay Details
Gene Symbol: pgp-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077727.9 | NP_510128.1 | 1 - 2 | 102 | 74 | |
GenBank mRNA | AY519858.1 | - | 1 - 2 | 91 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
