Assay Details
| Gene Symbol: pgp-14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077727.9 | NP_510128.1 | 2 - 3 | 316 | 99 | |
| GenBank mRNA | AY519858.1 | - | 2 - 3 | 305 | 99 | |
More Information
Set Membership: |
|

pgp-14
Genomic Map