Assay Details
| Gene Symbol: gei-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083303.5 | NP_001076772.1 | 18 - 19 | 2361 | 65 | |
| NM_001083304.6 | NP_001076773.1 | 1 - 2 | 66 | 65 | ||
More Information
Set Membership: |
|

gei-3
Genomic Map