Assay Details
| Gene Symbol: gei-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083303.5 | NP_001076772.1 | 21 - 22 | 2806 | 87 | |
| NM_001083304.6 | NP_001076773.1 | 4 - 5 | 511 | 87 | ||
More Information
Set Membership: |
|

gei-3
Genomic Map