Assay Details
| Gene Symbol: gei-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083303.5 | NP_001076772.1 | 22 - 23 | 3108 | 85 | |
| NM_001083304.6 | NP_001076773.1 | 5 - 6 | 813 | 85 | ||
More Information
Set Membership: |
|

gei-3
Genomic Map