Assay Details
| Gene Symbol: gei-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083303.5 | NP_001076772.1 | 27 - 28 | 3696 | 65 | |
| NM_001083304.6 | NP_001076773.1 | 10 - 11 | 1401 | 65 | ||
| NM_001083305.4 | NP_001076774.1 | 3 - 4 | 354 | 65 | ||
More Information
Set Membership: |
|

gei-3
Genomic Map