Assay Details
| Gene Symbol: gei-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083303.5 | NP_001076772.1 | 29 - 30 | 4115 | 75 | |
| NM_001083304.6 | NP_001076773.1 | 12 - 13 | 1820 | 75 | ||
| NM_001083305.4 | NP_001076774.1 | 5 - 6 | 773 | 75 | ||
More Information
Set Membership: |
|

gei-3
Genomic Map