Assay Details
| Gene Symbol: gei-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083303.5 | NP_001076772.1 | 3 - 4 | 252 | 105 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

gei-3| Gene Symbol: gei-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083303.5 | NP_001076772.1 | 3 - 4 | 252 | 105 | |
Set Membership: |
|