Assay Details
| Gene Symbol: chd-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077739.7 | NP_510140.1 | 7 - 8 | 5306 | 74 | |
| GenBank mRNA | AF308444.1 | - | 7 - 8 | 5328 | 74 | |
More Information
Set Membership: |
|

chd-3
Genomic Map