Assay Details
| Gene Symbol: bcat-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077743.5 | NP_510144.1 | 5 - 6 | 1225 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

bcat-1| Gene Symbol: bcat-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077743.5 | NP_510144.1 | 5 - 6 | 1225 | 97 | |
Set Membership: |
|