Assay Details
Gene Symbol: ppk-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077754.9 | NP_510155.3 | 12 - 13 | 3321 | 154 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
