Assay Details
| Gene Symbol: vwa-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077759.7 | NP_510160.2 | 23 - 24 | 4691 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

vwa-8| Gene Symbol: vwa-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077759.7 | NP_510160.2 | 23 - 24 | 4691 | 88 | |
Set Membership: |
|