Assay Details
| Gene Symbol: gck-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373560.4 | NP_001360654.1 | 2 | 374 | 92 | |
| NM_077889.4 | NP_510290.1 | 2 - 3 | 391 | 92 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

gck-4| Gene Symbol: gck-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373560.4 | NP_001360654.1 | 2 | 374 | 92 | |
| NM_077889.4 | NP_510290.1 | 2 - 3 | 391 | 92 | ||
Set Membership: |
|