Assay Details
| Gene Symbol: gck-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373560.4 | NP_001360654.1 | 8 | 3125 | 87 | |
| NM_077889.4 | NP_510290.1 | 8 - 9 | 3142 | 87 | ||
More Information
Set Membership: |
|

gck-4
Genomic Map