Assay Details
| Gene Symbol: poml-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077895.4 | NP_510296.2 | 4 - 5 | 469 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

poml-1| Gene Symbol: poml-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077895.4 | NP_510296.2 | 4 - 5 | 469 | 81 | |
Set Membership: |
|