Assay Details
Gene Symbol: pix-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270298.4 | NP_001257227.1 | 5 - 6 | 487 | 113 | |
NM_001270299.3 | NP_001257228.1 | 5 - 6 | 478 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
