Assay Details
| Gene Symbol: sra-38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077913.2 | NP_510314.1 | 2 - 3 | 242 | 160 | |
 Back To Top
 Back To TopMore Information
| Set Membership: |  Amplicon length greater than or equal to 101  Detect Genomic DNA  Caenorhabditis elegans  Probe spans exons | 
Gene Ontology Categories:
 Back To Top
 Back To Top
 sra-38
sra-38 Genomic Map
Genomic Map Back To Top
 Back To Top