Assay Details
Gene Symbol: alg-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381140.1 | NP_001367206.1 | 3 - 4 | 442 | 84 | |
NM_001381141.1 | NP_001366765.1 | 3 - 4 | 821 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
