Assay Details
Gene Symbol: del-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001368544.2 | NP_001355359.1 | 5 - 6 | 584 | 75 | |
NM_001368545.3 | NP_001355470.1 | 6 - 7 | 728 | 75 | ||
NM_001368546.2 | NP_001355471.1 | 5 - 6 | 533 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
