Assay Details
| Gene Symbol: C11H1.9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029220.4 | NP_001024391.1 | 4 - 5 | 667 | 108 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C11H1.9| Gene Symbol: C11H1.9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029220.4 | NP_001024391.1 | 4 - 5 | 667 | 108 | |
Set Membership: |
|