Assay Details
| Gene Symbol: ssq-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069367.5 | NP_501768.1 | 1 - 2 | 306 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ssq-1| Gene Symbol: ssq-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069367.5 | NP_501768.1 | 1 - 2 | 306 | 97 | |
Set Membership: |
|