Assay Details
| Gene Symbol: ctl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064578.5 | NP_496979.2 | 4 | 1477 | 120 | |
| Gene Symbol: ctl-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171051.4 | NP_741058.1 | 5 | 1551 | 120 | |
More Information
Set Membership: |
|

ctl-1
Genomic Map