Assay Details
Gene Symbol: cst-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383560.1 | 9 | 1216 | 78 |
Gene Symbol: C24A8.6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001361797.2 | NP_001348795.1 | 9 | 1224 | 78 | |
NM_001383559.1 | 10 | 1135 | 78 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001361797.2, NM_001383559.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
