Assay Details
| Gene Symbol: vps-41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380962.1 | NP_001367568.1 | 1 | 163 | 66 | |
| NM_001383575.1 | 1 | 168 | 66 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383575.1 transcripts.
Set Membership: |
|

vps-41
Genomic Map