Assay Details
Gene Symbol: ins-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268487.3 | NP_001255416.1 | 1 - 2 | 234 | 73 | |
NR_131589.1 | 2 - 3 | 281 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
