Assay Details
| Gene Symbol: C40H1.9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026043.4 | NP_001021214.2 | 2 - 3 | 258 | 114 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C40H1.9| Gene Symbol: C40H1.9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026043.4 | NP_001021214.2 | 2 - 3 | 258 | 114 | |
Set Membership: |
|