Assay Details
| Gene Symbol: Y18H1A.14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026562.4 | NP_001021733.2 | 2 - 3 | 593 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Y18H1A.14| Gene Symbol: Y18H1A.14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026562.4 | NP_001021733.2 | 2 - 3 | 593 | 67 | |
Set Membership: |
|