Assay Details
Gene Symbol: ins-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026792.5 | NP_001021963.1 | 2 - 3 | 264 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ins-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026792.5 | NP_001021963.1 | 2 - 3 | 264 | 83 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |