Assay Details
Gene Symbol: srz-67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026794.1 | NP_001021965.1 | 3 - 4 | 546 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: srz-67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026794.1 | NP_001021965.1 | 3 - 4 | 546 | 114 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |