Assay Details
| Gene Symbol: fut-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027139.2 | NP_001022310.1 | 6 - 7 | 933 | 123 | |
| GenBank mRNA | AJ745073.1 | - | 6 - 7 | 849 | 123 | |
More Information
Set Membership: |
|

fut-5
Genomic Map