Assay Details
Gene Symbol: srz-63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027312.1 | NP_001022483.1 | 4 - 5 | 688 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: srz-63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027312.1 | NP_001022483.1 | 4 - 5 | 688 | 124 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |