Assay Details
| Gene Symbol: hal-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001379881.2 | NP_001367068.1 | 14 - 15 | 1851 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

hal-2| Gene Symbol: hal-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001379881.2 | NP_001367068.1 | 14 - 15 | 1851 | 81 | |
Set Membership: |
|