Assay Details
| Gene Symbol: dlc-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028260.4 | NP_001023431.2 | 1 - 2 | 143 | 76 | |
| Gene Symbol: dlc-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083249.4 | NP_001076718.1 | 1 - 2 | 152 | 76 | |
More Information
Set Membership: |
|

dlc-4
Genomic Map