Assay Details
Gene Symbol: eff-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026816.6 | NP_001021987.1 | 2 - 3 | 351 | 162 | |
NM_001026817.5 | NP_001021988.1 | 2 - 3 | 357 | 162 | ||
NR_136365.1 | 2 - 3 | 347 | 162 | |||
GenBank mRNA | AF480492.1 | - | 2 - 3 | 347 | 162 | |
AF480493.1 | - | 2 - 3 | 347 | 162 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
