Assay Details
| Gene Symbol: W09G3.8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383751.1 | 10 - 11 | 3185 | 94 | ||
| Gene Symbol: maea-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061015.7 | NP_493416.1 | 2 - 3 | 206 | 94 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_061015.7 transcripts.
Set Membership: |
|

W09G3.8
Genomic Map