Assay Details
| Gene Symbol: glrx-22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038302.2 | NP_001033391.1 | 2 - 3 | 177 | 123 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

glrx-22| Gene Symbol: glrx-22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038302.2 | NP_001033391.1 | 2 - 3 | 177 | 123 | |
Set Membership: |
|