Assay Details
| Gene Symbol: aqp-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073111.7 | NP_505512.3 | 5 - 6 | 943 | 117 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

aqp-4| Gene Symbol: aqp-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073111.7 | NP_505512.3 | 5 - 6 | 943 | 117 | |
Set Membership: |
|