Assay Details
Gene Symbol: B0035.18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026090.2 | NP_001021261.1 | 2 | 248 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: B0035.18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026090.2 | NP_001021261.1 | 2 | 248 | 110 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |