Assay Details
| Gene Symbol: nlp-28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_071709.6 | NP_504110.1 | 1 - 2 | 122 | 158 | |
| Gene Symbol: nlp-27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_071710.8 | NP_504111.1 | 1 - 2 | 112 | 158 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_071710.8 transcripts.
Set Membership: |
|

nlp-28
Genomic Map