Assay Details
| Gene Symbol: SAMD4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005623414.3 | XP_005623471.1 | 7 - 8 | 1683 | 120 | |
| XM_005623415.3 | XP_005623472.1 | 8 - 9 | 1795 | 120 | ||
| XM_005623416.3 | XP_005623473.1 | 7 - 8 | 1634 | 120 | ||
| XM_014115912.2 | XP_013971387.1 | 3 - 4 | 556 | 120 | ||
| XM_859342.5 | XP_864435.2 | 7 - 8 | 2025 | 120 | ||
| XM_859395.5 | XP_864488.3 | 6 - 7 | 1761 | 120 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SAMD4A
Genomic Map