Assay Details
Gene Symbol: NBL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014111014.2 | XP_013966489.1 | 2 - 3 | 223 | 57 | |
XM_022413944.1 | XP_022269652.1 | 2 - 3 | 333 | 57 | ||
XM_022413951.1 | XP_022269659.1 | 2 - 3 | 328 | 57 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
