Assay Details
| Gene Symbol: OAZ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289914.1 | NP_001276843.1 | 3 - 4 | 579 | 61 | |
| GenBank mRNA | DN425590.1 | - | 3 - 4 | 579 | 61 | |
| DN864348.1 | - | 2 - 3 | 188 | 61 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

OAZ2
Genomic Map