Assay Details
Gene Symbol: AMMECR1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005631861.3 | XP_005631918.1 | 7 - 8 | 1306 | 70 | |
XM_005631862.3 | XP_005631919.1 | 7 - 8 | 993 | 70 | ||
XM_005631863.3 | XP_005631920.1 | 6 - 7 | 973 | 70 | ||
XM_014121461.2 | XP_013976936.1 | 6 - 7 | 995 | 70 | ||
XM_022406007.1 | XP_022261715.1 | 6 - 7 | 1184 | 70 | ||
XM_843911.5 | XP_849004.1 | 6 - 7 | 1175 | 70 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
