Assay Details
| Gene Symbol: SAMD11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005620343.3 | XP_005620400.1 | 1735 | 76 | ||
| XM_005620344.3 | XP_005620401.2 | 1755 | 76 | |||
| XM_005620345.2 | XP_005620402.1 | 1127 | 76 | |||
| XM_005620346.3 | XP_005620403.1 | 1110 | 76 | |||
| XM_014113787.2 | XP_013969262.1 | 1741 | 76 | |||
| XM_014113788.2 | XP_013969263.1 | 1730 | 76 | |||
| XM_014113790.2 | XP_013969265.1 | 1124 | 76 | |||
| XM_536715.4 | XP_536715.2 | 1112 | 76 | |||
More Information
Set Membership: |
|

SAMD11
Genomic Map