Assay Details
| Gene Symbol: AQP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003130.1 | NP_001003130.1 | 2 - 3 | 588 | 66 | |
| GenBank mRNA | AB011373.1 | - | 2 - 3 | 588 | 66 | |
| AB011374.1 | - | 1 - 2 | 128 | 66 | ||
| AF180528.1 | - | 2 - 3 | 236 | 66 | ||
More Information
Set Membership: |
|

AQP1
Genomic Map